Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RGSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137669.1 | NP_001131141.1 | 4 | 232 | ||
XM_005245137.1 | XP_005245194.1 | 4 | 217 | |||
XM_011509494.2 | XP_011507796.1 | 4 | 217 | |||
XM_011509496.1 | XP_011507798.1 | 4 | 217 | |||
XM_011509500.1 | XP_011507802.1 | 4 | 217 | |||
XM_011509503.1 | XP_011507805.1 | 4 | 217 | |||
XM_011509504.2 | XP_011507806.1 | 4 | 217 | |||
XM_017001188.1 | XP_016856677.1 | 4 | 217 | |||
XM_017001189.1 | XP_016856678.1 | 4 | 248 | |||
XM_017001190.1 | XP_016856679.1 | 4 | 1028 | |||
XM_017001191.1 | XP_016856680.1 | 4 | 229 | |||
XM_017001192.1 | XP_016856681.1 | 4 | 518 | |||
XM_017001193.1 | XP_016856682.1 | 4 | 345 | |||
XM_017001194.1 | XP_016856683.1 | 4 | 232 | |||
XM_017001195.1 | XP_016856684.1 | 3 | 174 | |||
XM_017001197.1 | XP_016856686.1 | 3 | 174 | |||
GenBank mRNA | AK302768.1 | - | 4 | 207 | ||
AL136902.1 | - | 3 | 196 | |||
BC142944.1 | - | 4 | 207 | 8860326 | ||
DB097250.1 | - | 4 | 232 |