Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요? 계정 생성하기
  • 모든 제품 주문
    • 항체
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR 분석
    • 피펫 및 피펫 팁
    • 실험실용 원심분리기
    • 초저온 냉동고
    • 분광학
    • 비이커
    • PCR 장비 및 용품
    • 제품 카테고리 전체보기
  • 응용 분야 및 기법
    • 세포 분석
    • 실험실 장비
    • Real-Time PCR
    • PCR
    • 크로마토그래피
    • 세포 배양 및 트랜스펙션
    • DNA/RNA 추출과 분석
    • 단백질 생물학
    • 유세포분석
    • 화학 제품
    • 응용 분야 및 기법 전체보기
  • 서비스
    • 맞춤형 서비스
    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo

Search

전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPB8 SIRNA Assays ›
          Gene Symbol:
          NPIPB8 LOC613037 NPIPA1 NPIPA2 NPIPA3 NPIPA5 NPIPA7 NPIPA8 NPIPB15 NPIPB3 NPIPB4 NPIPB5 NPIPB6 NPIPB9 PDXDC2P PKD1P1 PKD1P6-NPIPP1
          Gene Name:
          nuclear pore complex interacting protein family member B8
          nuclear pore complex interacting protein member
          nuclear pore complex interacting protein family member A1
          nuclear pore complex interacting protein family member A2
          nuclear pore complex interacting protein family member A3
          nuclear pore complex interacting protein family member A5
          nuclear pore complex interacting protein family member A7
          nuclear pore complex interacting protein family member A8
          nuclear pore complex interacting protein family member B15
          nuclear pore complex interacting protein family member B3
          nuclear pore complex interacting protein family member B4
          nuclear pore complex interacting protein family member B5
          nuclear pore complex interacting protein family member B6
          nuclear pore complex interacting protein family member B9
          pyridoxal dependent decarboxylase domain containing 2, pseudogene
          polycystin 1, transient receptor potential channel interacting pseudogene 1
          PKD1P6-NPIPP1 readthrough
          Gene Aliases:
          -
          -
          NPIP, NPIPA, morpheus
          NPIP
          -
          NPIP
          -
          LCR16a9
          A-761H5.4, NPIPL2
          KIAA0220L, NPIP, NPIPB, NPIPB5, NPIPL3
          61E3.4
          -
          NPIPB
          -
          NPIP, PDXDC2
          HG1, NPIP
          -
          Chromosome Location:
          Chr.16: 28638065 - 28658693 on Build GRCh38
          Chromosome Location:
          Chr.16: 30222735 - 30251993 on Build GRCh38
          Chromosome Location:
          Chr.16: 14935258 - 14952074 on Build GRCh38
          Chromosome Location:
          Chr.16: 14745595 - 14765448 on Build GRCh38
          Chromosome Location:
          Chr.16: 14706473 - 14726338 on Build GRCh38
          Chromosome Location:
          Chr.16: 15363624 - 15381478 on Build GRCh38
          Chromosome Location:
          Chr.16: 16376940 - 16393972 on Build GRCh38
          Chromosome Location:
          Chr.16: 18317919 - 18334967 on Build GRCh38
          Chromosome Location:
          Chr.16: 74374986 - 74392115 on Build GRCh38
          Chromosome Location:
          Chr.16: 21402134 - 21425337 on Build GRCh38
          Chromosome Location:
          Chr.16: 21834563 - 21859614 on Build GRCh38
          Chromosome Location:
          Chr.16: 22513523 - 22536521 on Build GRCh38
          Chromosome Location:
          Chr.16: 28342517 - 28364971 on Build GRCh38
          Chromosome Location:
          Chr.16: 28735500 - 28772911 on Build GRCh38
          Chromosome Location:
          Chr.16: 69976297 - 70065948 on Build GRCh38
          Chromosome Location:
          Chr.16: 16317609 - 16350608 on Build GRCh38
          Chromosome Location:
          Chr.16: 15104723 - 15131601 on Build GRCh38
          UniGene ID:
          -
          Hs.585733
          Hs.626677
          -
          -
          Hs.676266
          Hs.634612
          Hs.636569
          -
          Hs.611072
          Hs.611072
          Hs.611072
          Hs.726467
          Hs.710214
          Hs.513695
          Hs.636569
          Hs.634612
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 249599

          Purification
          Size
          재고 정보 본사 재고 보유
          Catalog # AM16708
          제품 정가 4,126.00
          Your Price
          온라인 행사가:
          4126.0
          공급가 확인 ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPB8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310136.1 NP_001297065.1 7 813
          XM_017023622.1 XP_016879111.1 8 1084
          XM_017023623.1 XP_016879112.1 8 1084
          XM_017023624.1 XP_016879113.1 8 937
          XM_017023625.1 XP_016879114.1 8 917
          XM_017023626.1 XP_016879115.1 9 1556
          Gene Symbol: LOC613037
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321892.1 NP_001308821.1 9 952
          XM_017023514.1 XP_016879003.1 9 1241
          XM_017023515.1 XP_016879004.1 10 1260
          XM_017023516.1 XP_016879005.1 9 1287
          XM_017023517.1 XP_016879006.1 9 1018
          XM_017023518.1 XP_016879007.1 9 1106
          XM_017023519.1 XP_016879008.1 9 1163
          XM_017023520.1 XP_016879009.1 4 2227
          XM_017023521.1 XP_016879010.1 5 482
          XM_017023522.1 XP_016879011.1 2 891
          Gene Symbol: NPIPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006985.2 NP_008916.2 8 759
          XM_011522748.2 XP_011521050.1 8 856
          XM_011522749.2 XP_011521051.1 9 968
          XM_011522750.2 XP_011521052.1 7 774
          XM_011522751.1 XP_011521053.1 9 925
          XM_017023891.1 XP_016879380.1 10 1038
          GenBank mRNA AF132984.1 - 8 759
          AK160377.1 - 28 5016
          CR627265.1 - 8034
          KF175225.1 - 8 759
          Gene Symbol: NPIPA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277324.1 NP_001264253.1 8 759
          XM_005255489.3 XP_005255546.1 10 3178
          Gene Symbol: NPIPA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277323.1 NP_001264252.1 8 759
          XM_005255484.3 XP_005255541.1 9 847
          XM_011522576.2 XP_011520878.2 10 3212
          XM_011522577.2 XP_011520879.2 10 3133
          XM_011522578.2 XP_011520880.2 8 855
          XM_011522580.2 XP_011520882.2 8 786
          XM_017023553.1 XP_016879042.1 10 3222
          XM_017023554.1 XP_016879043.1 9 3135
          Gene Symbol: NPIPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277325.1 NP_001264254.1 8 759
          XM_011522336.2 XP_011520638.1 11 1233
          Gene Symbol: NPIPA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282507.1 NP_001269436.1 8 759
          XM_005255735.4 XP_005255792.1 8 881
          XM_011522339.2 XP_011520641.1 8 807
          XM_017022833.1 XP_016878322.1 9 832
          XM_017022834.1 XP_016878323.1 9 884
          XM_017022835.1 XP_016878324.1 9 945
          XM_017022836.1 XP_016878325.1 8 752
          XM_017022837.1 XP_016878326.1 9 1695
          XM_017022838.1 XP_016878327.1 8 2090
          XM_017022839.1 XP_016878328.1 8 1587
          GenBank mRNA BI046418.1 - 5 410
          BI832721.1 - 2 141
          Gene Symbol: NPIPA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282511.1 NP_001269440.1 8 759
          XM_005255738.4 XP_005255795.1 8 881
          XM_006720830.3 XP_006720893.1 8 2642
          XM_011522342.2 XP_011520644.1 8 808
          XM_017022841.1 XP_016878330.1 9 832
          XM_017022842.1 XP_016878331.1 9 884
          XM_017022843.1 XP_016878332.1 9 945
          XM_017022844.1 XP_016878333.1 8 752
          XM_017022845.1 XP_016878334.1 9 1695
          XM_017022846.1 XP_016878335.1 8 1587
          Gene Symbol: NPIPB15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306094.1 NP_001293023.1 7 781
          XM_011523496.2 XP_011521798.1 8 837
          XM_011523498.2 XP_011521800.1 8 871
          XM_011523501.2 XP_011521803.1 4 663
          XM_017023248.1 XP_016878737.1 8 1274
          XM_017023249.1 XP_016878738.1 7 1091
          Gene Symbol: NPIPB3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_130464.2 NP_569731.2 8 835
          GenBank mRNA AB209632.1 - 7 3402
          AK128772.1 - 3 1955
          AK294452.1 - 4 398
          AK295170.1 - 8 834
          AK300277.1 - 152
          AK303166.1 - 1 42
          AK303500.1 - 4 329
          AK304162.1 - 6 617
          AK316459.1 - 4 264
          BC036263.1 - 8 835 3903371
          BC061522.1 - 1 143 6190707
          BC094882.1 - 7 762 3093162
          BM557638.1 - 4 409
          BQ721066.1 - 1 142
          KF175223.1 - 7 759
          KF175224.1 - 7 702
          Gene Symbol: NPIPB4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310148.1 NP_001297077.1 8 984
          XM_017023244.1 XP_016878733.1 7 2995
          XM_017023245.1 XP_016878734.1 8 2832
          XM_017023246.1 XP_016878735.1 8 2753
          XM_017023247.1 XP_016878736.1 7 986
          Gene Symbol: NPIPB5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135865.1 NP_001129337.1 7 1038
          GenBank mRNA AK296301.1 - 319
          AK296338.1 - 7 879
          AK303264.1 - 4 351
          AK303572.1 - 1 420
          Gene Symbol: NPIPB6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282524.1 NP_001269453.1 7 851
          XM_005255741.3 XP_005255798.1 8 1663
          XM_005255743.3 XP_005255800.1 4 2242
          XM_011545929.2 XP_011544231.1 9 1483
          XM_011545931.2 XP_011544233.1 8 1116
          XM_017023628.1 XP_016879117.1 8 866
          XM_017023629.1 XP_016879118.1 8 1409
          XM_017023630.1 XP_016879119.1 9 1136
          XM_017023631.1 XP_016879120.1 7 1480
          XM_017023632.1 XP_016879121.1 8 866
          GenBank mRNA BC144439.1 - 7 812
          BC157836.1 - 7 1501 9057055
          KF175222.1 - 7 813
          Gene Symbol: NPIPB9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287250.1 NP_001274179.1 7 851
          NM_001287251.1 NP_001274180.1 8 937
          XM_003118719.7 XP_003118767.2 8 1412
          XM_011545690.2 XP_011543992.1 10 1573
          XM_011545693.2 XP_011543995.1 9 1500
          XM_017022814.1 XP_016878303.1 9 1648
          XM_017022815.1 XP_016878304.1 9 1168
          XM_017022816.1 XP_016878305.1 9 1648
          XM_017022817.1 XP_016878306.1 8 1560
          XM_017022818.1 XP_016878307.1 8 2885
          XM_017022819.1 XP_016878308.1 9 1575
          XM_017022820.1 XP_016878309.1 9 1292
          XM_017022821.1 XP_016878310.1 8 2158
          XM_017022822.1 XP_016878311.1 9 3161
          XM_017022823.1 XP_016878312.1 7 2797
          XM_017022824.1 XP_016878313.1 9 1427
          XM_017022825.1 XP_016878314.1 9 1145
          XM_017022826.1 XP_016878315.1 8 1109
          XM_017022827.1 XP_016878316.1 4 856
          GenBank mRNA BC171825.1 - 8 1587
          Gene Symbol: PDXDC2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003610.1 26 3865
          GenBank mRNA AL833251.1 - 26 3865
          BC063633.1 - 24 2352
          BX647358.1 - 2 5429
          Gene Symbol: PKD1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036447.1 31 5895
          GenBank mRNA AF229069.1 - 1 118
          AK124516.1 - 26 4150
          BC039707.1 - 10 1148 4479933
          BC046145.1 - 10 1170
          Gene Symbol: PKD1P6-NPIPP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_123721.1 28 4922
          GenBank mRNA AK131084.1 - 23 5286
          AK131085.1 - 22 5192

          Back To Top

          추가 정보


          Additional Information:

          This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          mRNA transport
          carboxylic acid metabolic process
          protein binding
          carboxy-lyase activity
          pyridoxal phosphate binding

          Back To Top

          관련 제품

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          About Thermo Fisher Plus Icon Minus Icon
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)

           

          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)

           

          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0