Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CRYGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017541.2 | NP_060011.1 | 2 | 252 | ||
GenBank mRNA | AF161703.1 | - | 2 | 244 | ||
AK314172.1 | - | 2 | 252 | |||
AL133079.1 | - | 1 | 1102 | |||
BC069478.1 | - | 2 | 250 | 7262271 | ||
BC070241.1 | - | 3 | 540 | 30405628 | ||
KT966393.1 | - | 2 | 240 | |||
L36869.1 | - | 1 | 143 |