Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OBSCN SIRNA Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
UniGene ID:
Hs.650039
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
251364
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OBSCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098623.2
NP_001092093.2
11
3310
NM_001271223.2
NP_001258152.2
12
3586
NM_052843.3
NP_443075.3
11
3310
XM_005273287.4
XP_005273344.1
12
3666
XM_005273291.4
XP_005273348.1
12
3666
XM_005273298.4
XP_005273355.1
12
3666
XM_005273307.4
XP_005273364.1
12
3666
XM_006711822.3
XP_006711885.1
12
3666
XM_006711823.3
XP_006711886.1
12
3666
XM_006711827.3
XP_006711890.1
12
3665
XM_006711829.3
XP_006711892.1
11
3389
XM_011544297.2
XP_011542599.1
12
3666
XM_011544299.2
XP_011542601.1
11
3390
XM_017002443.1
XP_016857932.1
12
3666
XM_017002444.1
XP_016857933.1
12
3629
XM_017002445.1
XP_016857934.1
12
3666
XM_017002446.1
XP_016857935.1
12
3666
XM_017002447.1
XP_016857936.1
12
3666
XM_017002448.1
XP_016857937.1
12
3516
XM_017002449.1
XP_016857938.1
12
3667
XM_017002450.1
XP_016857939.1
12
3667
XM_017002451.1
XP_016857940.1
12
3667
XM_017002452.1
XP_016857941.1
12
3667
XM_017002453.1
XP_016857942.1
12
3666
XM_017002454.1
XP_016857943.1
11
3390
XM_017002455.1
XP_016857944.1
12
3666
XM_017002456.1
XP_016857945.1
11
3390
XM_017002457.1
XP_016857946.1
12
3666
XM_017002458.1
XP_016857947.1
12
3667
XM_017002459.1
XP_016857948.1
12
3667
XM_017002460.1
XP_016857949.1
12
3667
XM_017002461.1
XP_016857950.1
12
3666
XM_017002462.1
XP_016857951.1
10
3114
XM_017002463.1
XP_016857952.1
12
3666
XM_017002464.1
XP_016857953.1
12
3666
XM_017002465.1
XP_016857954.1
12
3667
XM_017002466.1
XP_016857955.1
12
3667
XM_017002467.1
XP_016857956.1
12
3667
XM_017002468.1
XP_016857957.1
12
3667
XM_017002469.1
XP_016857958.1
12
3667
XM_017002470.1
XP_016857959.1
12
3668
XM_017002471.1
XP_016857960.1
12
3665
XM_017002472.1
XP_016857961.1
12
3665
XM_017002473.1
XP_016857962.1
12
3665
GenBank mRNA
AJ002535.1
-
11
3307
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-blthd:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline