Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요?
계정 생성하기
모든 제품 주문
항체
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR 분석
피펫 및 피펫 팁
실험실용 원심분리기
초저온 냉동고
분광학
비이커
PCR 장비 및 용품
제품 카테고리 전체보기
응용 분야 및 기법
세포 분석
실험실 장비
Real-Time PCR
PCR
크로마토그래피
세포 배양 및 트랜스펙션
DNA/RNA 추출과 분석
단백질 생물학
유세포분석
화학 제품
응용 분야 및 기법 전체보기
서비스
맞춤형 서비스
엔터프라이즈급 실험실 정보 처리
엔터프라이즈 서비스
CDMO 및 임상 시험 서비스
장비 서비스
교육 서비스
서비스 전체 보기
지원
고객센터
문의하기
시험성적서(COA) 및 적합성 인증서(COC)
Safety Data Sheets (SDS)
매뉴얼
인용 및 참고 문헌
기기 지원
기술 자료/제품 FAQ
학습 센터
지원 메뉴 전체보기
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
문의하기
빠른 주문
주문 현황
제품 문서 검색
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
전체검색
전체검색
Search button
Search button
Close
주문 현황
빠른주문
로그인
로그인
회원이 아니신가요?
계정 생성하기
계정정보
주문내역조회
커스텀제품 및 프로젝트
장비관리
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HCLS1 SIRNA Assays ›
Gene Symbol:
Hcls1
Gene Name:
hematopoietic cell specific Lyn substrate 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 66736966 - 66759402 on Build Rnor_6.0
UniGene ID:
Rn.15578
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
251990
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hcls1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011898.1
NP_001011898.1
10
756
GenBank mRNA
BC079081.1
-
10
756
7122756
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
response to hormone
positive regulation of phosphatidylinositol 3-kinase signaling
actin filament polymerization
erythrocyte differentiation
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of tyrosine phosphorylation of STAT protein
positive regulation of transcription factor import into nucleus
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
negative regulation of leukocyte apoptotic process
RNA polymerase II transcription factor binding
actin binding
SH3 domain binding
protein kinase binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-swfkw:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0