Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024764.1 | NP_001019935.1 | 2 | 231 | ||
XM_006246381.2 | XP_006246443.1 | 2 | 274 | |||
XM_008767685.1 | XP_008765907.1 | 3 | 211 | |||
GenBank mRNA | BC097463.1 | - | 2 | 231 | 7457579 |