Search
Search

MMACHC| Gene Symbol: MMACHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015506.2 | NP_056321.2 | 4 | 2188 | ||
| XM_005270724.4 | XP_005270781.1 | 3 | 1947 | |||
| XM_011541204.2 | XP_011539506.1 | 4 | 2097 | |||
| GenBank mRNA | AL080062.1 | - | 5 | 1580 | ||