Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TARP SIRNA Assays ›
          Gene Symbol:
          TARP
          Gene Name:
          TCR gamma alternate reading frame protein
          Gene Aliases:
          CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
          Chromosome Location:
          Chr.7: 38239577 - 38276741 on Build GRCh38
          UniGene ID:
          Hs.534032
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 254225

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TARP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003799.1 NP_001003799.1 1 14
          NM_001003806.1 NP_001003806.1 1 14
          XM_011515407.2 XP_011513709.1 1 12
          GenBank mRNA AF151103.1 - 1 14
          AF512449.1 - 2 317
          AF512451.1 - 2 317
          AF512455.1 - 2 317
          AF512457.1 - 2 317
          JQ956403.1 - 2 156
          X72500.1 - 3 380
          Z11169.1 - 2 103
          Z11176.1 - 2 100
          Z11179.1 - 2 106
          Z11182.1 - 2 103
          Z11183.1 - 1, 2 106
          Z11184.1 - 2 106
          Z11187.1 - 2 100
          Z11188.1 - 2 103
          Z11190.1 - 2 103
          Z11192.1 - 2 103
          Z11197.1 - 2 99
          Z11202.1 - 2 103
          Z11204.1 - 1, 2 106
          Z11211.1 - 2 103
          Z11223.1 - 2 109
          Z11224.1 - 2 103
          Z11225.1 - 2 106
          Z11236.1 - 2 109
          Z11239.1 - 2 103
          Z11252.1 - 2 103
          Z11253.1 - 2 106
          Z11256.1 - 2 115
          Z11257.1 - 2 103
          Z11272.1 - 2 103
          Z11274.1 - 2 106
          Z11275.1 - 2 103
          Z11278.1 - 2 106
          Z11279.1 - 2 103
          Z11281.1 - 2 106
          Z11283.1 - 2 103
          Z11284.1 - 2 103
          Z11285.1 - 2 103
          Z11287.1 - 2 106
          Z11288.1 - 1, 2 106
          Z11291.1 - 2 106
          Z11294.1 - 2 103
          Z11297.1 - 2 103
          Z11301.1 - 2 103
          Z11302.1 - 2 103
          Z11307.1 - 2 100
          Z11308.1 - 2 109
          Z11319.1 - 1, 2 103
          Z11321.1 - 2 103
          Z11326.1 - 2 103
          Z11327.1 - 1, 2 106
          Z11333.1 - 2 103
          Z11334.1 - 2 103
          Z11335.1 - 2 103
          Z11341.1 - 2 106
          Z11348.1 - 2 103
          Z11350.1 - 2 106
          Z11354.1 - 2 103
          Z11357.1 - 2 102
          Z11360.1 - 2 103
          Z11362.1 - 2 106
          Z11363.1 - 2 103
          Z11367.1 - 2 103
          Z11369.1 - 2 106
          Z11373.1 - 2 103
          Z11374.1 - 2 100
          Z11387.1 - 2 109
          Z11388.1 - 2, 3 103
          Z11390.1 - 2 106
          Z11394.1 - 2 103
          Z11397.1 - 2 103
          Z11408.1 - 2 112
          Z11410.1 - 1, 2 106

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0