Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP1 SIRNA Assays ›
          Gene Symbol:
          Rplp1 RGD1564744
          Gene Name:
          ribosomal protein, large, P1
          similar to 60S acidic ribosomal protein P1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 18613039 - 18613557 on Build Rnor_6.0
          UniGene ID:
          Rn.973
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 254572

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 3 461
          GenBank mRNA BC058151.1 - 3 464 6888011
          FQ209966.1 - 3 463
          FQ210237.1 - 3 460
          FQ210375.1 - 3 460
          FQ210407.1 - 3 460
          FQ210632.1 - 3 462
          FQ210864.1 - 3 460
          FQ210988.1 - 3 463
          FQ211097.1 - 3 463
          FQ211327.1 - 3 461
          FQ211648.1 - 3 460
          FQ212351.1 - 3 460
          FQ214973.1 - 3 462
          FQ215065.1 - 3 460
          FQ216905.1 - 3 464
          FQ216921.1 - 3 463
          FQ216959.1 - 3 459
          FQ216979.1 - 3 463
          FQ217216.1 - 3 463
          FQ217313.1 - 3 461
          FQ220167.1 - 3 461
          FQ220212.1 - 3 462
          FQ220998.1 - 3 460
          FQ221027.1 - 3 461
          FQ221120.1 - 3 461
          FQ221137.1 - 3 460
          FQ221201.1 - 3 463
          FQ221359.1 - 3 461
          FQ221455.1 - 3 465
          FQ221567.1 - 3 463
          FQ221595.1 - 3 461
          FQ221620.1 - 3 460
          FQ221698.1 - 3 467
          FQ221717.1 - 3 460
          FQ221756.1 - 3 460
          FQ221784.1 - 3 460
          FQ221894.1 - 3 460
          FQ221969.1 - 3 460
          FQ221980.1 - 3 460
          FQ222031.1 - 3 460
          FQ222178.1 - 3 460
          FQ222307.1 - 3 462
          FQ222468.1 - 3 462
          FQ222559.1 - 3 460
          FQ222618.1 - 3 463
          FQ222784.1 - 3 460
          FQ222801.1 - 3 461
          FQ222805.1 - 3 460
          FQ222816.1 - 3 461
          FQ222923.1 - 3 461
          FQ222965.1 - 3 460
          FQ222966.1 - 3 459
          FQ223013.1 - 3 461
          FQ223016.1 - 3 462
          FQ223066.1 - 3 462
          FQ223337.1 - 3 460
          FQ223344.1 - 3 461
          FQ223388.1 - 3 460
          FQ223390.1 - 3 461
          FQ223470.1 - 3 463
          FQ223512.1 - 3 461
          FQ223537.1 - 3 461
          FQ223567.1 - 3 460
          FQ223591.1 - 3 460
          FQ223674.1 - 3 442
          FQ223769.1 - 3 461
          FQ224482.1 - 3 430
          FQ224625.1 - 3 463
          FQ224766.1 - 3 461
          FQ228323.1 - 3 460
          FQ228361.1 - 3 460
          FQ228390.1 - 3 461
          FQ228490.1 - 3 461
          FQ228528.1 - 3 457
          FQ228539.1 - 3 460
          FQ228605.1 - 3 464
          FQ228650.1 - 3 460
          FQ228990.1 - 3 467
          FQ229009.1 - 3 460
          FQ229050.1 - 3 460
          FQ229318.1 - 3 461
          FQ230089.1 - 3 461
          X15097.1 - 3 431
          Gene Symbol: RGD1564744
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006241133.2 XP_006241195.1 2 429
          XM_006241134.2 XP_006241196.1 2 423
          XM_006241135.2 XP_006241197.1 2 423
          XM_008765205.1 XP_008763427.1 2 429
          XM_234961.5 XP_234961.1 1 483

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: BC172075, BC157910, BC158067.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0