Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GOLGA8N SIRNA Assays ›
          Gene Symbol:
          GOLGA8N GOLGA8CP GOLGA8DP GOLGA8EP GOLGA8H GOLGA8J GOLGA8M GOLGA8O GOLGA8R LOC101930434
          Gene Name:
          golgin A8 family member N
          golgin A8 family member C, pseudogene
          golgin A8 family member D, pseudogene
          golgin A8 family member E, pseudogene
          golgin A8 family member H
          golgin A8 family member J
          golgin A8 family member M
          golgin A8 family member O
          golgin A8 family member R
          putative golgin subfamily A member 8I
          Gene Aliases:
          GOLGA8R
          GOLGA8C
          GOLGA8D
          GOLGA8E
          GOLGA6L11
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.15: 32593456 - 32607310 on Build GRCh38
          Chromosome Location:
          Chr.15: 20562349 - 20575703 on Build GRCh38
          Chromosome Location:
          Chr.15: 23157340 - 23170783 on Build GRCh38
          Chromosome Location:
          Chr.15: 22424673 - 22438026 on Build GRCh38
          Chromosome Location:
          Chr.15: 30604030 - 30617827 on Build GRCh38
          Chromosome Location:
          Chr.15: 30082627 - 30096711 on Build GRCh38
          Chromosome Location:
          Chr.15: 28698695 - 28718512 on Build GRCh38
          Chromosome Location:
          Chr.15: 32441914 - 32456323 on Build GRCh38
          Chromosome Location:
          Chr.15: 30400562 - 30414260 on Build GRCh38
          Chromosome Location:
          Chr.15: 30798436 - 30802763 on Build GRCh38
          UniGene ID:
          Hs.672754
          Hs.592025
          Hs.454647
          Hs.454647
          -
          Hs.574257
          Hs.562625
          Hs.525905
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 255167

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GOLGA8N
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282494.1 NP_001269423.1 14, 15 1382
          Gene Symbol: GOLGA8CP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027411.2 13, 14 1696
          Gene Symbol: GOLGA8DP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027407.1 13, 14 1684
          GenBank mRNA AK292534.1 - 13, 14 1567
          BC110417.1 - 13, 14 1655 5171794
          Gene Symbol: GOLGA8EP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033350.1 13, 14 1684
          GenBank mRNA BC063309.1 - 14, 15 1658
          DQ309037.1 - 13, 14 1658
          Gene Symbol: GOLGA8H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282490.1 NP_001269419.1 14, 15 1380
          Gene Symbol: GOLGA8J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282472.1 NP_001269401.1 14, 15 1382
          XM_006720651.3 XP_006720714.1 14, 15 1402
          XM_011521948.2 XP_011520250.1 14, 15 1710
          Gene Symbol: GOLGA8M
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282468.1 NP_001269397.1 14, 15 1382
          XM_005268284.4 XP_005268341.1 14, 15 2181
          XM_005268285.4 XP_005268342.1 13, 14 2112
          XM_006720654.3 XP_006720717.1 14, 15 1325
          XM_011521951.2 XP_011520253.1 15, 16 1750
          XM_011521952.2 XP_011520254.1 14, 15 2181
          XM_011521954.2 XP_011520256.1 14, 15 1315
          Gene Symbol: GOLGA8O
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277308.1 NP_001264237.1 14, 15 1382
          XM_011521988.2 XP_011520290.1 13, 14 1998
          Gene Symbol: GOLGA8R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282484.1 NP_001269413.1 14, 15 1379
          Gene Symbol: LOC101930434
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017022809.1 XP_016878298.1 3, 4 178

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0