Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdsub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014179.1 | NP_001014201.1 | 8 | 1139 | ||
XM_006234208.2 | XP_006234270.1 | 8 | 1399 | |||
XM_006234209.2 | XP_006234271.1 | 8 | 1398 | |||
XM_006234211.2 | XP_006234273.1 | 8 | 1120 | |||
GenBank mRNA | BC089856.1 | - | 8 | 1139 | 7381399 |