Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
UniGene ID:
Mm.138471
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
256516
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Eef1a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010106.2
NP_034236.2
4
625
GenBank mRNA
AK011771.1
-
4
573
AK012341.1
-
4
573
AK032914.1
-
4
571
AK050635.1
-
4
572
AK076696.1
-
4
573
AK081725.1
-
4
573
AK083361.1
-
4
573
AK088041.1
-
4
574
AK133725.1
-
4
573
AK135448.1
-
4
574
AK137223.1
-
4
573
AK141243.1
-
4
575
AK150798.1
-
4
573
AK151479.1
-
4
574
AK151773.1
-
4
573
AK152810.1
-
4
573
AK152993.1
-
4
574
AK153387.1
-
4
573
AK153402.1
-
4
573
AK153851.1
-
4
573
AK162049.1
-
4
573
AK162313.1
-
4
572
AK162440.1
-
4
577
AK163799.1
-
4
573
AK164113.1
-
4
575
AK164671.1
-
4
573
AK165938.1
-
4
573
AK166021.1
-
4
573
AK166033.1
-
4
573
AK166058.1
-
4
573
AK166089.1
-
4
573
AK166615.1
-
4
573
AK166807.1
-
4
573
AK167843.1
-
4
573
AK167873.1
-
4
572
AK168424.1
-
4
573
AK168467.1
-
4
579
AK187140.1
-
1
106
AK188863.1
-
2
267
AK206299.1
-
1
106
AK209036.1
-
1
104
AK210348.1
-
1
107
AK213893.1
-
1
106
BC003969.1
-
3
543
BC004005.1
-
3
534
3492770
BC004067.1
-
3
562
3590965
BC005660.1
-
3
563
3588966
BC018223.1
-
4
561
4012653
BC018485.1
-
4
575
3491827
BC040689.1
-
1
65
BC050124.1
-
2118
BC068258.1
-
2370
BC083069.1
-
4
561
6310448
BC092053.1
-
3
551
4008574
BC092276.1
-
3
556
3490007
BC092292.1
-
3
517
BC095965.1
-
3
551
6310404
BC108391.1
-
4
558
30242277
M22432.1
-
4
564
X13661.1
-
3
544
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_175838, BC111707, L10339, X61043, X63561, and others.
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0