Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012508.3 | NP_001012526.2 | 2 | 2147 | ||
GenBank mRNA | AK123191.1 | - | 1 | 804 | ||
BC040198.1 | - | 2 | 2109 | 4816858 |