Search
Search

YOD1| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276320.1 | NP_001263249.1 | 4 | 5006 | ||
| NM_018566.3 | NP_061036.3 | 2 | 4980 | |||
| GenBank mRNA | AL833081.2 | - | 2 | 4913 | ||
| BC137166.1 | - | 2 | 4973 | 9021163 | ||
| BC137167.1 | - | 2 | 4974 | 9021164 | ||