Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POU5F1 SIRNA Assays ›
Gene Symbol:
Pou5f1
Gene Name:
POU class 5 homeobox 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 3747231 - 3751994 on Build Rnor_6.0
UniGene ID:
Rn.161748
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
258419
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pou5f1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009178.2
NP_001009178.1
5
933
GenBank mRNA
BC158566.1
-
5
933
9092936
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mesodermal cell fate commitment
endodermal cell fate commitment
ectodermal cell fate commitment
endodermal cell fate specification
trophectodermal cell differentiation
blastocyst growth
BMP signaling pathway involved in heart induction
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to wounding
regulation of asymmetric cell division
response to organic substance
regulation of gene expression
stem cell population maintenance
germ-line stem cell population maintenance
response to retinoic acid
response to cytokine
somatic stem cell population maintenance
positive regulation of catenin import into nucleus
mRNA transcription from RNA polymerase II promoter
cell fate commitment
negative regulation of cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of calcium ion-dependent exocytosis
stem cell differentiation
positive regulation of protein kinase B signaling
negative regulation of protein kinase B signaling
positive regulation of SMAD protein import into nucleus
cell fate commitment involved in formation of primary germ layer
cardiac cell fate determination
negative regulation of gene silencing by miRNA
regulation of heart induction by regulation of canonical Wnt signaling pathway
regulation of methylation-dependent chromatin silencing
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription coactivator activity
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
transcription factor binding
ubiquitin protein ligase binding
miRNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
poly(A) RNA binding
protein heterodimerization activity
POU domain binding
HMG box domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-kzg96:80/100.66.78.128:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0