Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIMS2 SIRNA Assays ›
          Gene Symbol:
          LIMS2
          Gene Name:
          LIM zinc finger domain containing 2
          Gene Aliases:
          LGMD2W, PINCH-2, PINCH2
          Chromosome Location:
          Chr.2: 127638421 - 127681786 on Build GRCh38
          UniGene ID:
          Hs.469881
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 25885

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIMS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136037.2 NP_001129509.2 11 1242
          NM_001161403.1 NP_001154875.1 10 1176
          NM_001161404.1 NP_001154876.1 10 1466
          NM_001256542.1 NP_001243471.1 7 868
          NM_017980.4 NP_060450.2 10 1287
          XM_005263710.2 XP_005263767.1 9 959
          XM_006712627.3 XP_006712690.1 7 768
          XM_011511453.1 XP_011509755.1 9 1023
          XM_017004469.1 XP_016859958.1 5 733
          GenBank mRNA AF484961.1 - 9 1010
          AF520987.1 - 10 1630
          AF527764.1 - 10 1241
          AF527765.1 - 10 1103
          AF527766.1 - 10 1143
          AF527767.1 - 9 1399
          AF527768.1 - 8 758
          AF527769.1 - 6 832
          AF527770.1 - 7 795
          AK000906.1 - 8 658
          AK022470.1 - 10 1055
          AK055363.1 - 10 1631
          AK091652.1 - 8 1537
          AK093692.1 - 4 1230
          AK094954.1 - 9 1723
          AK095790.1 - 13 1691
          AK123014.1 - 5 3935
          AK297645.1 - 12 1434
          AL512716.1 - 1 115
          BC001370.2 - 2 266
          BC008977.1 - 2 181
          BC065816.1 - 10 1094 6181259
          BX458594.2 - 7 824

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell junction protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single organismal cell-cell adhesion
          cell junction assembly
          negative regulation of apoptotic process
          cell-cell junction organization
          negative regulation of neural precursor cell proliferation
          negative regulation of hepatocyte proliferation
          positive regulation of integrin-mediated signaling pathway
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline