Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
25893
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
11
1406
NR_047653.1
11
1467
NR_047654.1
11
1467
NR_047655.1
10
1315
NR_047656.1
10
1315
NR_047657.1
11
1406
NR_047658.1
11
1383
XM_005263982.2
XP_005264039.1
11
1405
XM_005263984.1
XP_005264041.1
11
1405
XM_005263987.1
XP_005264044.1
11
1405
XM_011511454.2
XP_011509756.1
11
1405
XM_011511455.2
XP_011509757.1
11
1374
XM_011511456.2
XP_011509758.1
11
1408
XM_011511457.2
XP_011509759.1
11
1327
XM_011511458.2
XP_011509760.1
11
1405
XM_011511459.2
XP_011509761.1
10
1330
XM_011511460.2
XP_011509762.1
11
1405
XM_011511461.2
XP_011509763.1
11
1405
XM_011511462.2
XP_011509764.1
10
1234
XM_011511463.2
XP_011509765.1
11
1418
XM_011511464.2
XP_011509766.1
11
1477
XM_011511466.2
XP_011509768.1
11
1418
XM_011511467.2
XP_011509769.1
10
1260
XM_017004470.1
XP_016859959.1
11
1405
XM_017004471.1
XP_016859960.1
10
1312
XM_017004472.1
XP_016859961.1
11
1418
XM_017004473.1
XP_016859962.1
11
1418
XM_017004474.1
XP_016859963.1
11
1313
GenBank mRNA
AB037731.1
-
11
1358
AF311326.1
-
13
1219
AK000943.1
-
9
1307
AK023813.1
-
11
1434
AK302646.1
-
10
1256
AK304011.1
-
10
1315
AK311617.1
-
10
1248
AK314367.1
-
11
1343
AY050169.1
-
11
1434
BC015469.2
-
12
1400
BC024276.1
-
11
1343
BC032746.1
-
11
1477
BC051763.1
-
9
1037
BC063792.1
-
11
1336
6504387
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mkfsp:80/100.66.78.65:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0