Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요? 계정 생성하기
  • 모든 제품 주문
    • 항체
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR 분석
    • 피펫 및 피펫 팁
    • 실험실용 원심분리기
    • 초저온 냉동고
    • 분광학
    • 비이커
    • PCR 장비 및 용품
    • 제품 카테고리 전체보기
  • 응용 분야 및 기법
    • 세포 분석
    • 실험실 장비
    • Real-Time PCR
    • PCR
    • 크로마토그래피
    • 세포 배양 및 트랜스펙션
    • DNA/RNA 추출과 분석
    • 단백질 생물학
    • 유세포분석
    • 화학 제품
    • 응용 분야 및 기법 전체보기
  • 서비스
    • 맞춤형 서비스
    • 엔터프라이즈급 실험실 정보 처리
    • 엔터프라이즈 서비스
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상 시험 CRO 서비
    • 장비 서비스
    • 교육 서비스
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo

Search

전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other USP17L6P SIRNA Assays ›
          Gene Symbol:
          USP17L6P USP17L10 USP17L11 USP17L12 USP17L13 USP17L15 USP17L17 USP17L18 USP17L19 USP17L2 USP17L20 USP17L21 USP17L22 USP17L24 USP17L25 USP17L26 USP17L27 USP17L28 USP17L29 USP17L30 USP17L5 USP17L9P
          Gene Name:
          ubiquitin specific peptidase 17-like family member 6, pseudogene
          ubiquitin specific peptidase 17-like family member 10
          ubiquitin specific peptidase 17-like family member 11
          ubiquitin specific peptidase 17-like family member 12
          ubiquitin specific peptidase 17-like family member 13
          ubiquitin specific peptidase 17-like family member 15
          ubiquitin specific peptidase 17-like family member 17
          ubiquitin specific peptidase 17-like family member 18
          ubiquitin specific peptidase 17-like family member 19
          ubiquitin specific peptidase 17-like family member 2
          ubiquitin specific peptidase 17-like family member 20
          ubiquitin specific peptidase 17-like family member 21
          ubiquitin specific peptidase 17-like family member 22
          ubiquitin specific peptidase 17-like family member 24
          ubiquitin specific peptidase 17-like family member 25
          ubiquitin specific peptidase 17-like family member 26
          ubiquitin specific peptidase 17-like family member 27
          ubiquitin specific peptidase 17-like family member 28
          ubiquitin specific peptidase 17-like family member 29
          ubiquitin specific peptidase 17-like family member 30
          ubiquitin specific peptidase 17-like family member 5
          ubiquitin specific peptidase 17-like family member 9, pseudogene
          Gene Aliases:
          DUB4, USP17L6, vDUB4
          -
          -
          -
          -
          -
          -
          -
          -
          DUB-3, DUB3, USP17
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          RS447, USP17, USP17A, USP17H, USP17I, USP17J, USP17K, USP17L, USP17M
          Chromosome Location:
          Chr.4: 9367874 - 9369070 on Build GRCh38
          Chromosome Location:
          Chr.4: 9210657 - 9212249 on Build GRCh38
          Chromosome Location:
          Chr.4: 9215405 - 9216997 on Build GRCh38
          Chromosome Location:
          Chr.4: 9220152 - 9221744 on Build GRCh38
          Chromosome Location:
          Chr.4: 9224896 - 9226488 on Build GRCh38
          Chromosome Location:
          Chr.4: 9234385 - 9236334 on Build GRCh38
          Chromosome Location:
          Chr.4: 9243879 - 9245471 on Build GRCh38
          Chromosome Location:
          Chr.4: 9248630 - 9250222 on Build GRCh38
          Chromosome Location:
          Chr.4: 9253378 - 9254970 on Build GRCh38
          Chromosome Location:
          Chr.8: 12137168 - 12138760 on Build GRCh38
          Chromosome Location:
          Chr.4: 9258124 - 9259716 on Build GRCh38
          Chromosome Location:
          Chr.4: 9262872 - 9264464 on Build GRCh38
          Chromosome Location:
          Chr.4: 9267619 - 9269211 on Build GRCh38
          Chromosome Location:
          Chr.4: 9325165 - 9326757 on Build GRCh38
          Chromosome Location:
          Chr.4: 9329911 - 9331503 on Build GRCh38
          Chromosome Location:
          Chr.4: 9334658 - 9336250 on Build GRCh38
          Chromosome Location:
          Chr.4: 9344148 - 9345740 on Build GRCh38
          Chromosome Location:
          Chr.4: 9348893 - 9350485 on Build GRCh38
          Chromosome Location:
          Chr.4: 9353638 - 9355230 on Build GRCh38
          Chromosome Location:
          Chr.4: 9363129 - 9364721 on Build GRCh38
          Chromosome Location:
          Chr.4: 9339403 - 9340995 on Build GRCh38
          Chromosome Location:
          Chr.4: 9358382 - 9359974 on Build GRCh38
          UniGene ID:
          Hs.722658
          Hs.741130
          Hs.722783
          Hs.741128
          Hs.741127
          Hs.528621
          Hs.741129
          Hs.741138
          Hs.741137
          Hs.531448
          Hs.722783
          Hs.741136
          Hs.723053
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.553810
          Hs.740243
          Hs.553810
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 258984

          Purification
          Size
          재고 정보 본사 재고 보유
          Catalog # AM16708
          제품 정가 4,126.00
          Your Price
          온라인 행사가:
          4126.0
          공급가 확인 ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: USP17L6P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027279.1 1 1106
          GenBank mRNA AY533200.1 - 1 1106
          Gene Symbol: USP17L10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256852.1 NP_001243781.1 1 1106
          Gene Symbol: USP17L11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256854.1 NP_001243783.1 1 1106
          Gene Symbol: USP17L12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256853.1 NP_001243782.1 1 1106
          Gene Symbol: USP17L13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256855.1 NP_001243784.1 1 1106
          Gene Symbol: USP17L15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256894.1 NP_001243823.1 1 1106
          Gene Symbol: USP17L17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256857.1 NP_001243786.1 1 1106
          Gene Symbol: USP17L18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256859.1 NP_001243788.1 1 1106
          Gene Symbol: USP17L19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256860.1 NP_001243789.1 1 1106
          Gene Symbol: USP17L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY509884.1 - 1 1106
          Gene Symbol: USP17L20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256861.1 NP_001243790.1 1 1106
          GenBank mRNA AY188990.1 - 1 1106
          Gene Symbol: USP17L21
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256862.1 NP_001243791.1 1 1106
          Gene Symbol: USP17L22
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256863.1 NP_001243792.1 1 1106
          Gene Symbol: USP17L24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242327.1 NP_001229256.1 1 1106
          Gene Symbol: USP17L25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242326.1 NP_001229255.1 1 1106
          Gene Symbol: USP17L26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242328.1 NP_001229257.1 1 1106
          Gene Symbol: USP17L27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242330.1 NP_001229259.1 1 1106
          Gene Symbol: USP17L28
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242331.1 NP_001229260.1 1 1106
          Gene Symbol: USP17L29
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242332.1 NP_001229261.1 1 1106
          Gene Symbol: USP17L30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256867.1 NP_001243796.1 1 1106
          Gene Symbol: USP17L5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242329.1 NP_001229258.1 1 1106
          Gene Symbol: USP17L9P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046416.1 1 1107
          GenBank mRNA AF544011.1 - 2 1106
          AF544012.1 - 1 1106
          BC100991.2 - 1 1106
          BC100992.2 - 1 1106
          BC100993.1 - 1 1109

          Back To Top

          추가 정보


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          protein deubiquitination
          apoptotic process
          thiol-dependent ubiquitinyl hydrolase activity

          Back To Top

          관련 제품

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          About Thermo Fisher Plus Icon Minus Icon
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)

           

          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)

           

          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0