Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZCCHC11 SIRNA Assays ›
Gene Symbol:
ZCCHC11
Gene Name:
zinc finger CCHC-type containing 11
Gene Aliases:
PAPD3, TUT4
Chromosome Location:
Chr.1: 52423275 - 52553463 on Build GRCh38
UniGene ID:
Hs.655407
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
259168
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
283.00
Your Price
Online offer:
274.65
274.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZCCHC11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009881.2
NP_001009881.1
13
2327
NM_015269.2
NP_056084.1
13
2327
XM_005270676.2
XP_005270733.1
13
2336
XM_005270678.2
XP_005270735.1
13
2336
XM_005270679.2
XP_005270736.1
13
2336
XM_005270680.2
XP_005270737.1
13
2336
XM_005270681.2
XP_005270738.1
13
2336
XM_005270682.2
XP_005270739.1
11
2003
XM_005270683.2
XP_005270740.1
13
2336
XM_006710498.2
XP_006710561.1
13
2273
XM_006710499.3
XP_006710562.1
13
2280
XM_006710500.1
XP_006710563.1
12
2123
XM_011541097.1
XP_011539399.1
14
2358
XM_011541098.1
XP_011539400.1
13
2255
XM_011541099.1
XP_011539401.1
12
2042
XM_011541100.2
XP_011539402.1
13
2336
XM_011541101.2
XP_011539403.1
13
2336
XM_017000803.1
XP_016856292.1
13
2255
XM_017000804.1
XP_016856293.1
13
2255
XM_017000805.1
XP_016856294.1
13
2336
XM_017000806.1
XP_016856295.1
12
2123
XM_017000807.1
XP_016856296.1
12
2123
XM_017000808.1
XP_016856297.1
11
2003
XM_017000809.1
XP_016856298.1
11
2003
XM_017000810.1
XP_016856299.1
11
2003
XM_017000811.1
XP_016856300.1
13
2336
XM_017000812.1
XP_016856301.1
13
2336
XM_017000813.1
XP_016856302.1
13
2336
XM_017000814.1
XP_016856303.1
11
2003
XM_017000815.1
XP_016856304.1
13
2336
XM_017000816.1
XP_016856305.1
13
2255
XM_017000817.1
XP_016856306.1
12
2123
XM_017000818.1
XP_016856307.1
11
2003
GenBank mRNA
AK307579.1
-
13
2336
AK308427.1
-
13
1542
BC131734.1
-
13
2219
40125005
BX648783.1
-
12
1724
D83776.1
-
12
1692
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
miRNA metabolic process
miRNA catabolic process
stem cell population maintenance
pre-miRNA processing
RNA 3'-end processing
histone mRNA catabolic process
protein binding
zinc ion binding
poly(A) RNA binding
RNA uridylyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline