Search
Search

IWS1| Gene Symbol: IWS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017969.2 | NP_060439.2 | 10, 11 | 2346 | ||
| XM_005263707.3 | XP_005263764.1 | 10, 11 | 2182 | |||
| XM_006712626.3 | XP_006712689.1 | 10, 11 | 2260 | |||
| XM_011511450.1 | XP_011509752.1 | 10, 11 | 2260 | |||
| XM_017004467.1 | XP_016859956.1 | 10, 11 | 2347 | |||
| XM_017004468.1 | XP_016859957.1 | 10, 11 | 2116 | |||
| GenBank mRNA | AK000868.1 | - | 10, 11 | 1673 | ||
| AK001717.1 | - | 5, 6 | 591 | |||
| AK027561.1 | - | 10, 11 | 2303 | |||
| AK056881.1 | - | 10, 11 | 1347 | |||
| AK294669.1 | - | 11, 12 | 2054 | |||
| AK296848.1 | - | 11, 12 | 2154 | |||
| AL834178.1 | - | 10, 11 | 2294 | |||
| BC017012.2 | - | 7, 8 | 823 | |||
| BC110536.1 | - | 10, 11 | 2176 | 40035173 | ||
| BC110537.1 | - | 10, 11 | 2176 | 40035174 | ||