Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013176.1 | NP_001013194.1 | 4 | 758 | ||
XM_006243035.2 | XP_006243097.1 | 6 | 1515 | |||
XM_006243036.2 | XP_006243098.1 | 5 | 915 | |||
XM_006243038.2 | XP_006243100.1 | 5 | 812 | |||
XM_008766262.1 | XP_008764484.1 | 5 | 949 | |||
GenBank mRNA | BC088755.1 | - | 4 | 758 | 7307514 |