Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002875.1 | - | 4 | 452 | ||
AK142609.1 | - | 4 | 594 | |||
BC003495.1 | - | 5 | 624 | 2812450 | ||
BC003844.1 | - | 4 | 620 | 3497357 | ||
BC004786.1 | - | 4 | 640 | 3588636 | ||
BC010324.1 | - | 6 | 636 | 2651406 | ||
BC010798.1 | - | 5 | 561 | |||
BC011181.1 | - | 4 | 584 | 4207118 | ||
BC013488.1 | - | 5 | 584 | 4215488 | ||
BC013490.1 | - | 6 | 587 | 4216550 | ||
BC013539.1 | - | 5 | 617 | 4241603 | ||
BC013656.1 | - | 4 | 628 | 4209551 | ||
BC018322.1 | - | 4 | 605 | 4208089 | ||
BC018461.1 | - | 4 | 638 | |||
BC019425.1 | - | 4 | 631 | 4459683 | ||
BC019569.1 | - | 4 | 625 | 4037595 | ||
BC031703.1 | - | 4 | 604 | 4205959 | ||
BC049143.1 | - | 4 | 638 | 6474993 | ||
BC092056.1 | - | 4 | 616 | 4013094 | ||
BC103785.1 | - | 3 | 608 | 6477061 | ||
BC110689.1 | - | 4 | 606 | 4221167 |
Gene Symbol: Igha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136471.1 | - | 641 | |||
BC055919.1 | - | 604 |