Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014616.2 | NP_055431.1 | 19 | 2442 | ||
XM_005247241.2 | XP_005247298.1 | 19 | 2441 | |||
XM_005247243.2 | XP_005247300.1 | 18 | 2113 | |||
XM_011512593.2 | XP_011510895.1 | 19 | 2441 | |||
XM_011512594.2 | XP_011510896.1 | 19 | 2441 | |||
XM_011512597.2 | XP_011510899.1 | 17 | 2008 | |||
XM_017006008.1 | XP_016861497.1 | 17 | 2005 | |||
XM_017006009.1 | XP_016861498.1 | 19 | 2211 | |||
GenBank mRNA | AB023173.1 | - | 6 | 672 | ||
AK093727.1 | - | 7 | 914 | |||
AK296750.1 | - | 9 | 1031 | |||
AK304855.1 | - | 9 | 1335 | |||
BC010630.1 | - | 5 | 570 |