Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRUNE2 SIRNA Assays ›
Gene Symbol:
PRUNE2
Gene Name:
prune homolog 2
Gene Aliases:
BMCC1, BNIPXL, C9orf65, KIAA0367
Chromosome Location:
Chr.9: 76611376 - 76906220 on Build GRCh38
UniGene ID:
Hs.262857
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
261165
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRUNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308047.1
NP_001294976.1
8
2401
NM_001308048.1
NP_001294977.1
8
2401
NM_015225.2
NP_056040.2
8
2401
NR_131751.1
7
2198
XM_005251745.1
XP_005251802.1
8
2534
XM_005251746.1
XP_005251803.1
8
2534
XM_005251748.1
XP_005251805.1
8
2534
XM_005251750.1
XP_005251807.1
8
2534
XM_005251751.1
XP_005251808.1
8
2534
XM_005251754.1
XP_005251811.1
8
2534
XM_006716982.1
XP_006717045.1
8
2534
XM_006716983.1
XP_006717046.1
8
2534
XM_006716984.2
XP_006717047.1
8
2534
XM_006716985.1
XP_006717048.1
8
2534
XM_006716986.1
XP_006717049.1
8
2534
XM_011518323.1
XP_011516625.1
8
2534
XM_011518326.2
XP_011516628.1
8
2534
XM_011518327.1
XP_011516629.1
8
2534
XM_011518328.2
XP_011516630.1
8
2534
XM_017014345.1
XP_016869834.1
8
2534
XM_017014346.1
XP_016869835.1
8
2534
XM_017014347.1
XP_016869836.1
8
2534
XM_017014348.1
XP_016869837.1
8
2534
XM_017014349.1
XP_016869838.1
8
2534
XM_017014350.1
XP_016869839.1
8
2534
XM_017014351.1
XP_016869840.1
8
2534
XM_017014352.1
XP_016869841.1
8
2534
XM_017014353.1
XP_016869842.1
8
2534
GenBank mRNA
AB050197.1
-
2
1522
AB535152.1
-
8
2277
AL834213.1
-
1
571
BC150641.1
-
2
1419
9057011
FJ808772.1
-
8
2355
FJ808773.1
-
7
2152
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
pyrophosphatase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline