Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zrsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017504.1 | NP_001017504.1 | 1 | 1212 | ||
XM_008770435.1 | XP_008768657.1 | 2 | 3131 | |||
XM_008770436.1 | XP_008768658.1 | 2 | 3055 | |||
GenBank mRNA | BC086322.1 | - | 1 | 1212 | 7301896 |