Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
262307
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204106.1
NP_001191035.1
2
309
NM_001204107.1
NP_001191036.1
2
309
NM_001204108.1
NP_001191037.1
2
309
NM_001204109.1
NP_001191038.1
2
309
NM_001204110.1
NP_001191039.1
2
309
NM_001204111.1
NP_001191040.1
2
309
NM_001204112.1
NP_001191041.1
2
309
NM_001204113.1
NP_001191042.1
2
309
NM_006538.4
NP_006529.1
2
309
NM_138621.4
NP_619527.1
2
309
NM_138622.3
NP_619528.1
2
309
NM_138623.3
NP_619529.1
2
309
NM_138624.3
NP_619530.1
2
309
NM_138625.3
NP_619531.1
2
309
NM_138626.3
NP_619532.1
2
309
NM_138627.3
NP_619533.1
2
309
NM_207002.3
NP_996885.1
2
309
NM_207003.2
NP_996886.1
2
309
XM_005263550.3
XP_005263607.1
2
1087
XM_005263551.2
XP_005263608.1
2
1087
XM_005263552.3
XP_005263609.1
2
1087
XM_005263553.2
XP_005263610.1
2
1087
XM_005263554.2
XP_005263611.1
2
1087
XM_005263555.3
XP_005263612.1
2
1087
XM_005263556.3
XP_005263613.1
2
309
XM_005263557.4
XP_005263614.1
2
386
XM_005263559.2
XP_005263616.1
2
1087
XM_005263561.3
XP_005263618.1
2
309
XM_011510461.2
XP_011508763.1
2
1087
XM_011510464.2
XP_011508766.1
2
1087
XM_017003101.1
XP_016858590.1
2
1087
XM_017003102.1
XP_016858591.1
2
309
GenBank mRNA
AB071195.1
-
1
21
AB071196.1
-
1
21
AB071197.1
-
1
21
AB071198.1
-
1
21
AB071199.1
-
1
21
AB071200.1
-
1
21
AF032457.1
-
1
21
AF032458.1
-
1
21
AK290377.1
-
2
309
AK291269.1
-
2
306
AY305714.1
-
1
21
AY305715.1
-
1
21
AY352518.1
-
1
21
AY423441.1
-
1
21
AY423442.1
-
1
21
AY423443.1
-
1
21
AY428962.1
-
1
21
BC033694.1
-
2
291
5213713
CN411912.1
-
2
278
DQ849200.1
-
1
21
DQ849201.1
-
1
21
DQ849202.1
-
1
21
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_207680, NM_001291016, NM_207681, NM_009754, NM_001284410, and others.
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0