Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L11 SIRNA Assays ›
Gene Symbol:
BCL2L11
Gene Name:
BCL2 like 11
Gene Aliases:
BAM, BIM, BOD
Chromosome Location:
Chr.2: 111120914 - 111168445 on Build GRCh38
UniGene ID:
Hs.469658
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
262308
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204106.1
NP_001191035.1
2
397
NM_001204107.1
NP_001191036.1
2
397
NM_001204108.1
NP_001191037.1
2
397
NM_001204109.1
NP_001191038.1
2
397
NM_001204110.1
NP_001191039.1
2
397
NM_001204111.1
NP_001191040.1
2
397
NM_001204112.1
NP_001191041.1
2
397
NM_001204113.1
NP_001191042.1
2
397
NM_006538.4
NP_006529.1
2
397
NM_138621.4
NP_619527.1
2
397
NM_138622.3
NP_619528.1
2
397
NM_138623.3
NP_619529.1
2
397
NM_138624.3
NP_619530.1
2
397
NM_138625.3
NP_619531.1
2
397
NM_138626.3
NP_619532.1
2
397
NM_138627.3
NP_619533.1
2
397
NM_207002.3
NP_996885.1
2
397
NM_207003.2
NP_996886.1
2
397
XM_005263550.3
XP_005263607.1
2
1175
XM_005263551.2
XP_005263608.1
2
1175
XM_005263552.3
XP_005263609.1
2
1175
XM_005263553.2
XP_005263610.1
2
1175
XM_005263554.2
XP_005263611.1
2
1175
XM_005263555.3
XP_005263612.1
2
1175
XM_005263556.3
XP_005263613.1
2
397
XM_005263557.4
XP_005263614.1
2
474
XM_005263559.2
XP_005263616.1
2
1175
XM_005263561.3
XP_005263618.1
2
397
XM_011510461.2
XP_011508763.1
2
1175
XM_011510464.2
XP_011508766.1
2
1175
XM_017003101.1
XP_016858590.1
2
1175
XM_017003102.1
XP_016858591.1
2
397
GenBank mRNA
AB071195.1
-
1
109
AB071196.1
-
1
109
AB071197.1
-
1
109
AB071198.1
-
1
109
AB071199.1
-
1
109
AB071200.1
-
1
109
AF032457.1
-
1
109
AF032458.1
-
1
109
AK290377.1
-
2
397
AK291269.1
-
2
394
AY305714.1
-
1
109
AY305715.1
-
1
109
AY352518.1
-
1
109
AY423441.1
-
1
109
AY423442.1
-
1
109
AY423443.1
-
1
109
AY428962.1
-
1
109
BC033694.1
-
2
379
5213713
CN411912.1
-
2
366
DQ849200.1
-
1
109
DQ849201.1
-
1
109
DQ849202.1
-
1
109
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
B cell homeostasis
B cell apoptotic process
kidney development
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
myeloid cell homeostasis
apoptotic process
meiosis I
cell-matrix adhesion
spermatogenesis
brain development
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
mammary gland development
positive regulation of protein homooligomerization
response to endoplasmic reticulum stress
tube formation
odontogenesis of dentin-containing tooth
T cell homeostasis
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of neuron apoptotic process
ear development
positive regulation of cell cycle
regulation of organ growth
developmental pigmentation
regulation of developmental pigmentation
spleen development
thymus development
post-embryonic organ morphogenesis
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
thymocyte apoptotic process
positive regulation of release of cytochrome c from mitochondria
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
apoptotic process involved in embryonic digit morphogenesis
positive regulation of IRE1-mediated unfolded protein response
positive regulation of fibroblast apoptotic process
positive regulation of intrinsic apoptotic signaling pathway
protein binding
microtubule binding
dynein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0