Search
Search

AURKC| Gene Symbol: AURKC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015878.1 | NP_001015878.1 | 7 | 1240 | ||
| NM_001015879.1 | NP_001015879.1 | 7 | 1099 | |||
| NM_003160.2 | NP_003151.2 | 7 | 1104 | |||
| GenBank mRNA | AF054621.1 | - | 7 | 1245 | ||
| AF059681.1 | - | 7 | 1299 | |||
| BX092186.1 | - | 2 | 378 | |||
| EU668345.1 | - | 7 | 1299 | |||