Search
Search

Olr1585
LOC102550614| Gene Symbol: Olr1585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001109.1 | NP_001001109.1 | 1 | 524 | ||
| Gene Symbol: LOC102550614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250349.2 | XP_006250411.1 | 2 | 344 | ||