Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009467.3 | NP_033493.3 | 6 | 1658 | ||
GenBank mRNA | AK136480.1 | - | 6 | 1659 | ||
X06358.1 | - | 6 | 1636 |