Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP1R9A SIRNA Assays ›
Gene Symbol:
PPP1R9A
Gene Name:
protein phosphatase 1 regulatory subunit 9A
Gene Aliases:
NRB1, NRBI, Neurabin-I
Chromosome Location:
Chr.7: 94907202 - 95296415 on Build GRCh38
UniGene ID:
Hs.21816
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
263432
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP1R9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166160.1
NP_001159632.1
20
6152
NM_001166161.1
NP_001159633.1
18
5852
NM_001166162.1
NP_001159634.1
17
5723
NM_001166163.1
NP_001159635.1
14
5234
NM_017650.2
NP_060120.2
16
5324
XM_011516380.1
XP_011514682.1
21
6339
XM_011516381.1
XP_011514683.1
21
6215
XM_011516382.2
XP_011514684.1
21
6228
XM_011516383.1
XP_011514685.1
21
6269
XM_011516389.1
XP_011514691.1
19
6086
XM_017012394.1
XP_016867883.1
21
6161
XM_017012395.1
XP_016867884.1
22
6125
XM_017012397.1
XP_016867886.1
21
6101
XM_017012398.1
XP_016867887.1
19
6098
XM_017012399.1
XP_016867888.1
20
6056
XM_017012400.1
XP_016867889.1
20
5996
XM_017012401.1
XP_016867890.1
20
5984
XM_017012403.1
XP_016867892.1
19
5918
XM_017012404.1
XP_016867893.1
18
5453
XM_017012405.1
XP_016867894.1
16
5448
XM_017012406.1
XP_016867895.1
16
5379
XM_017012407.1
XP_016867896.1
15
5300
XM_017012408.1
XP_016867897.1
19
4430
XM_017012410.1
XP_016867899.1
20
6218
XM_017012411.1
XP_016867900.1
14
8832
GenBank mRNA
AB033048.1
-
15
3976
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
aging
calcium-mediated signaling
regulation of actin filament polymerization
neuron projection development
positive regulation of protein kinase activity
regulation of filopodium assembly
negative regulation of stress fiber assembly
regulation of synapse structural plasticity
regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of dendritic spine development
regulation of dendritic spine morphogenesis
cellular response to toxic substance
negative regulation of long-term synaptic potentiation
positive regulation of long term synaptic depression
negative regulation of spontaneous neurotransmitter secretion
protein binding
protein C-terminus binding
protein phosphatase 1 binding
protein kinase binding
protein domain specific binding
protein homodimerization activity
ion channel binding
actin filament binding
GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0