Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARHGEF10L SIRNA Assays ›
          Gene Symbol:
          ARHGEF10L
          Gene Name:
          Rho guanine nucleotide exchange factor 10 like
          Gene Aliases:
          GrinchGEF
          Chromosome Location:
          Chr.1: 17520197 - 17697875 on Build GRCh38
          UniGene ID:
          Hs.443460
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 26377

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARHGEF10L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001011722.2 NP_001011722.2 16, 17 1854
          NM_001319837.1 NP_001306766.1 15, 16 1839
          NM_001319838.1 NP_001306767.1 7, 8 1078
          NM_001328124.1 NP_001315053.1 10, 11 1303
          NM_018125.3 NP_060595.3 18, 19 2087
          NR_137287.1 18, 19 2090
          NR_137288.1 17, 18 1970
          XM_005245923.2 XP_005245980.2 17, 18 1946
          XM_005245925.2 XP_005245982.2 17, 18 1934
          XM_005245927.2 XP_005245984.2 10, 11 1303
          XM_005245929.2 XP_005245986.2 9, 10 1288
          XM_006710728.1 XP_006710791.1 17, 18 2128
          XM_006710729.1 XP_006710792.1 17, 18 1943
          XM_006710731.1 XP_006710794.1 17, 18 2125
          XM_011541691.2 XP_011539993.1 16, 17 1943
          XM_011541692.2 XP_011539994.1 15, 16 1937
          XM_011541693.2 XP_011539995.1 15, 16 1928
          XM_011541695.2 XP_011539997.1 16, 17 1952
          XM_011541696.2 XP_011539998.1 15, 16 1937
          XM_017001617.1 XP_016857106.1 16, 17 1919
          XM_017001618.1 XP_016857107.1 11, 12 1221
          XM_017001619.1 XP_016857108.1 16, 17 1952
          XM_017001620.1 XP_016857109.1 15, 16 1937
          XM_017001621.1 XP_016857110.1 17, 18 2070
          XM_017001622.1 XP_016857111.1 16, 17 2055
          GenBank mRNA AB046846.1 - 18, 19 2087
          AK001383.1 - 4, 5 545
          AK092020.1 - 7, 8 1075
          AK126877.1 - 9, 10 2193
          AK300174.1 - 14, 15 1618
          AK300264.1 - 9, 10 1251
          AK311076.1 - 17, 18 1897
          BC065561.1 - 4, 5 589
          BC080596.1 - 7, 8 750
          BC117171.1 - 16, 17 1844 40125722
          BC143347.1 - 16, 17 1844 9051852
          DQ139833.1 - 15, 16 1796

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of Rho protein signal transduction
          positive regulation of GTPase activity
          Rho guanyl-nucleotide exchange factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0