Search
Search

CYLC1| Gene Symbol: CYLC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271680.1 | NP_001258609.1 | 4 | 246 | ||
| NM_021118.2 | NP_066941.1 | 5 | 1995 | |||
| XM_005262086.3 | XP_005262143.1 | 5 | 1992 | |||
| GenBank mRNA | BC126461.1 | - | 5 | 1959 | 8992177 | |
| BX110674.1 | - | 4 | 207 | |||
| Z22780.1 | - | 3 | 1780 | |||