Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26521
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
15
2416
NM_001292043.1
NP_001278972.1
17
2680
XM_005265301.1
XP_005265358.1
15
2454
XM_005265303.1
XP_005265360.1
17
2835
XM_011533920.1
XP_011532222.1
18
2930
XM_011533921.1
XP_011532223.1
17
2532
XM_011533922.1
XP_011532224.1
16
2493
XM_011533927.1
XP_011532229.1
18
2873
XM_011533928.1
XP_011532230.1
16
2436
XM_011533930.1
XP_011532232.1
18
2874
XM_011533932.1
XP_011532234.1
16
2641
XM_011533933.1
XP_011532235.1
14
2010
XM_017006767.1
XP_016862256.1
18
4592
XM_017006768.1
XP_016862257.1
16
4106
XM_017006769.1
XP_016862258.1
16
6209
XM_017006770.1
XP_016862259.1
17
6248
XM_017006771.1
XP_016862260.1
15
2454
XM_017006772.1
XP_016862261.1
16
6209
XM_017006773.1
XP_016862262.1
16
6152
XM_017006774.1
XP_016862263.1
15
2397
XM_017006775.1
XP_016862264.1
16
6152
XM_017006776.1
XP_016862265.1
17
2737
XM_017006777.1
XP_016862266.1
17
2680
XM_017006778.1
XP_016862267.1
18
2721
XM_017006779.1
XP_016862268.1
16
2698
XM_017006780.1
XP_016862269.1
17
2682
XM_017006781.1
XP_016862270.1
18
2721
XM_017006782.1
XP_016862271.1
17
6248
XM_017006783.1
XP_016862272.1
19
2946
XM_017006784.1
XP_016862273.1
17
6248
XM_017006785.1
XP_016862274.1
17
6248
XM_017006786.1
XP_016862275.1
17
6248
GenBank mRNA
AB051544.1
-
11
2210
AK001569.1
-
4
599
AK024384.1
-
3
510
AK091393.1
-
12
1719
AK092046.1
-
17
2635
AK127298.1
-
5
1303
AK289529.1
-
6
998
AL442073.1
-
2
396
BC020956.1
-
4
630
3851178
BX648380.1
-
7
1018
CR627408.1
-
14
2032
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0