Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080462.1 | NP_001073931.1 | 5 | 744 | ||
XM_011521497.1 | XP_011519799.1 | 5 | 745 | |||
XM_011521499.1 | XP_011519801.1 | 5 | 565 | |||
GenBank mRNA | AK131033.1 | - | 3 | 382 |