Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018270.5 | NP_060740.1 | 2 | 352 | ||
NR_136405.1 | 2 | 352 | ||||
GenBank mRNA | AB085682.1 | - | 2 | 252 | ||
AK001776.1 | - | 2 | 304 | |||
BC009889.2 | - | 2 | 271 | 4026378 | ||
BC014235.2 | - | 2 | 276 | 4555060 | ||
BC018841.2 | - | 2 | 271 | 3050866 | ||
BM920599.1 | - | 258 | ||||
BU902487.1 | - | 2 | 353 |