Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143958.1 | NP_001137430.1 | 3 | 643 | ||
NM_018247.3 | NP_060717.1 | 4 | 751 | |||
GenBank mRNA | AK001718.1 | - | 4 | 620 | ||
AK292823.1 | - | 3 | 508 | |||
AK293225.1 | - | 5 | 576 | |||
AK295567.1 | - | 5 | 572 | |||
AL832490.1 | - | 4 | 674 | |||
AL832815.1 | - | 4 | 613 | |||
BC009006.1 | - | 3 | 494 | 4154200 |