Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ECHDC2 SIRNA Assays ›
Gene Symbol:
ECHDC2
Gene Name:
enoyl-CoA hydratase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 52895910 - 52927212 on Build GRCh38
UniGene ID:
Hs.476319
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26686
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ECHDC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198961.1
NP_001185890.1
4
400
NM_001198962.1
NP_001185891.1
4
400
NM_001319958.1
NP_001306887.1
5
913
NM_018281.3
NP_060751.2
4
400
XM_011541709.2
XP_011540011.1
5
611
XM_011541710.2
XP_011540012.1
5
611
XM_011541711.2
XP_011540013.1
5
611
XM_011541712.2
XP_011540014.1
5
611
XM_011541713.2
XP_011540015.1
4
4388
XM_011541715.2
XP_011540017.1
5
836
XM_011541719.2
XP_011540021.1
5
1833
XM_011541720.2
XP_011540022.1
4
4157
XM_011541721.2
XP_011540023.1
6
850
XM_011541722.2
XP_011540024.1
5
468
XM_011541723.2
XP_011540025.1
5
568
XM_011541724.2
XP_011540026.1
5
611
XM_011541725.2
XP_011540027.1
5
611
XM_011541726.2
XP_011540028.1
4
4388
XM_011541727.2
XP_011540029.1
4
4157
XM_017001638.1
XP_016857127.1
6
2222
XM_017001639.1
XP_016857128.1
5
627
XM_017001640.1
XP_016857129.1
4
790
XM_017001641.1
XP_016857130.1
6
1991
XM_017001642.1
XP_016857131.1
4
4388
GenBank mRNA
AK001810.1
-
4
352
AK299834.1
-
5
569
AK300896.1
-
5
439
AK302285.1
-
7
979
AK307381.1
-
4
295
AK310650.1
-
5
532
AK315906.1
-
5
531
BC038389.1
-
5
831
BC044574.1
-
4
330
5314083
BC051887.1
-
4
388
CF127686.1
-
4
344
Back To Top
More Information
Panther Classification:
Molecular Function -
hydratase
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
enoyl-CoA hydratase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0