Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C22orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110522.1 | 3 | 1021 | |||
NR_110523.1 | 5 | 1492 | ||||
GenBank mRNA | BC033837.1 | - | 5 | 1465 |