Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HSBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136180.1 | NP_001129652.1 | 3 | 343 | ||
GenBank mRNA | AK001829.1 | - | 1 | 667 | ||
BC017654.1 | - | 3 | 271 | |||
BC040056.2 | - | 3 | 267 | |||
BC157848.1 | - | 3 | 265 | 9057067 |