Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
CDMO and Clinical Trial Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEC16A SIRNA Assays ›
Gene Symbol:
SEC16A
Gene Name:
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases:
KIAA0310, SEC16L, p250
Chromosome Location:
Chr.9: 136440096 - 136484552 on Build GRCh38
UniGene ID:
Hs.522500
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
272304
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEC16A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276418.1
NP_001263347.1
22, 23
6631
NM_014866.1
NP_055681.1
23, 24
6621
XM_005266134.2
XP_005266191.1
22, 23
6484
XM_011519246.2
XP_011517548.1
23, 24
6630
XM_011519247.1
XP_011517549.1
22, 23
6487
XM_011519248.2
XP_011517550.1
22, 23
6461
XM_011519249.1
XP_011517551.1
22, 23
6484
XM_011519250.2
XP_011517552.1
22, 23
6484
XM_011519251.1
XP_011517553.1
22, 23
6484
XM_011519252.2
XP_011517554.1
22, 23
6484
XM_011519253.1
XP_011517555.1
22, 23
6484
XM_011519254.1
XP_011517556.1
22, 23
6484
XM_011519255.2
XP_011517557.1
22, 23
6484
XM_011519256.1
XP_011517558.1
22, 23
6484
XM_011519257.1
XP_011517559.1
22, 23
6484
XM_011519258.1
XP_011517560.1
21, 22
6343
XM_011519259.1
XP_011517561.1
22, 23
6484
XM_011519260.2
XP_011517562.1
22, 23
6468
XM_011519261.1
XP_011517563.1
19, 20
6037
XM_011519262.1
XP_011517564.1
23, 24
5773
XM_011519263.2
XP_011517565.1
23, 24
5773
XM_017015363.1
XP_016870852.1
22, 23
6484
GenBank mRNA
AB002308.4
-
23, 24
6621
AB593115.1
-
22, 23
6631
AK129634.1
-
2, 3
234
BC001404.2
-
6, 7
700
BC008332.1
-
12, 13
1494
BC028183.1
-
18, 19
2488
BC098454.1
-
6, 7
784
BC125018.1
-
21, 22
5193
BC125019.1
-
22, 23
4482
DQ903855.1
-
21, 22
5778
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endoplasmic reticulum organization
protein transport
substantia nigra development
COPII vesicle coating
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-swfkw:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0