Search
Search

PROX2| Gene Symbol: PROX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080408.2 | NP_001073877.2 | 2 | 831 | ||
| NM_001243007.1 | NP_001229936.1 | 3 | 1512 | |||
| XM_005267543.3 | XP_005267600.1 | 6 | 2590 | |||
| GenBank mRNA | AK094068.1 | - | 2 | 515 | ||
| BC105720.1 | - | 3 | 1511 | |||
| BC105927.1 | - | 3 | 1511 | |||