Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080468.2 | NP_001073937.1 | 1 | 373 | ||
GenBank mRNA | AK130951.1 | - | 1 | 384 | ||
BC121075.2 | - | 1 | 377 | 40119665 | ||
BC121076.2 | - | 1 | 377 | 40119666 |