Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DAZAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018959.3 | NP_061832.2 | 4, 5 | 507 | ||
NM_170711.2 | NP_733829.1 | 4, 5 | 507 | |||
XM_005259534.3 | XP_005259591.1 | 4, 5 | 466 | |||
XM_005259536.3 | XP_005259593.1 | 4, 5 | 468 | |||
XM_011527904.2 | XP_011526206.1 | 4, 5 | 1772 | |||
XM_011527905.2 | XP_011526207.1 | 4, 5 | 1780 | |||
XM_011527908.2 | XP_011526210.1 | 4, 5 | 1779 | |||
XM_011527909.2 | XP_011526211.1 | 4, 5 | 1779 | |||
XM_011527910.2 | XP_011526212.1 | 4, 5 | 859 | |||
XM_017026581.1 | XP_016882070.1 | 4, 5 | 502 | |||
XM_017026582.1 | XP_016882071.1 | 4, 5 | 1795 | |||
GenBank mRNA | AF181719.1 | - | 4, 5 | 410 | ||
AK056850.1 | - | 2, 3 | 130 | |||
AK125032.1 | - | 3, 4 | 1803 | |||
BC012062.1 | - | 4, 5 | 375 | 4549444 | ||
BI909277.1 | - | 4, 5 | 491 | |||
KU178593.1 | - | 4, 5 | 302 |