Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDE7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018945.3 | NP_061818.1 | 9, 10 | 1106 | ||
XM_005266931.3 | XP_005266988.1 | 8, 9 | 3955 | |||
XM_011535755.2 | XP_011534057.1 | 10, 11 | 935 | |||
XM_017010741.1 | XP_016866230.1 | 8, 9 | 1741 | |||
GenBank mRNA | AB038040.1 | - | 9, 10 | 1108 | ||
AB209990.1 | - | 8, 9 | 1212 | |||
AJ251860.1 | - | 9, 10 | 1100 | |||
AK290582.1 | - | 9, 10 | 1139 | |||
BC075082.2 | - | 9, 10 | 829 | 30915597 | ||
BC075083.2 | - | 9, 10 | 833 | 30915385 | ||
EF108327.1 | - | 8, 9 | 1214 |