Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 8, 9 | 1698 | ||
NM_001184966.1 | NP_001171895.1 | 7, 8 | 1623 | |||
NM_019045.4 | NP_061918.3 | 8, 9 | 1698 | |||
XM_011531353.2 | XP_011529655.1 | 7, 8 | 1699 | |||
XM_017029600.1 | XP_016885089.1 | 7, 8 | 1631 | |||
GenBank mRNA | AK001978.1 | - | 2, 3 | 166 | ||
AK092077.1 | - | 5, 6 | 702 | |||
AK127556.1 | - | 8, 9 | 1692 | |||
AK299711.1 | - | 7, 8 | 1559 | |||
AK304432.1 | - | 8, 9 | 1225 | |||
AL833605.1 | - | 8, 9 | 1663 | |||
AL834210.1 | - | 5, 6 | 965 | |||
BC028697.2 | - | 8, 9 | 1634 | 4837674 | ||
BX537578.1 | - | 8, 9 | 1673 | |||
BX538087.1 | - | 8, 9 | 1640 | |||
BX648150.1 | - | 8, 9 | 1637 |