Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 3 | 361 | ||
XM_005268466.1 | XP_005268523.1 | 3 | 411 | |||
XM_005268467.1 | XP_005268524.1 | 3 | 411 | |||
XM_005268468.1 | XP_005268525.1 | 3 | 411 | |||
XM_006714787.1 | XP_006714850.1 | 3 | 411 | |||
XM_011537661.1 | XP_011535963.1 | 3 | 411 | |||
XM_011537662.1 | XP_011535964.1 | 3 | 411 | |||
XM_017009587.1 | XP_016865076.1 | 3 | 411 | |||
GenBank mRNA | AK128738.1 | - | 3 | 411 | ||
BC009537.1 | - | 3 | 364 | |||
BX647384.1 | - | 3 | 361 |