Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001483.2 | NP_001001483.1 | 16 | 1938 | ||
NM_019028.2 | NP_061901.2 | 17 | 2084 | |||
XM_005252996.2 | XP_005253053.1 | 17 | 2056 | |||
XM_011520193.1 | XP_011518495.1 | 14 | 1697 | |||
XM_011520195.1 | XP_011518497.1 | 17 | 1994 | |||
GenBank mRNA | AB024495.1 | - | 17 | 2032 | ||
AB097014.1 | - | 17 | 2082 | |||
AB097027.1 | - | 17 | 2082 | |||
AK001714.1 | - | 17 | 2082 | |||
AK001831.1 | - | 12 | 1456 | |||
AK123539.1 | - | 3 | 1785 | |||
AK291348.1 | - | 17 | 2059 | |||
AL833714.1 | - | 13 | 1534 | |||
BC036020.1 | - | 17 | 2069 | 5272445 | ||
BC050690.1 | - | 16 | 1938 | 6171322 | ||
BC056152.1 | - | 17 | 2084 | 6170701 | ||
BX648156.1 | - | 15 | 5950 |