Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019029.2 | NP_061902.2 | 12 | 1454 | ||
NM_031311.3 | NP_112601.3 | 12 | 1366 | |||
XM_011515437.1 | XP_011513739.1 | 12 | 1533 | |||
XM_017012366.1 | XP_016867855.1 | 10 | 1093 | |||
GenBank mRNA | AF106704.2 | - | 12 | 1286 | ||
AF217508.1 | - | 11 | 1254 | |||
AF282617.1 | - | 12 | 1316 | |||
AK075433.1 | - | 12 | 1367 | |||
AK124472.1 | - | 16 | 1894 | |||
AK225545.1 | - | 12 | 1317 | |||
AY358549.1 | - | 12 | 1343 | |||
BC016838.2 | - | 12 | 1414 | 3888647 |