Search
Search

MAP7D3| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173516.1 | NP_001166987.1 | 4 | 462 | ||
| NM_001173517.1 | NP_001166988.1 | 4 | 574 | |||
| NM_024597.3 | NP_078873.2 | 4 | 574 | |||
| XM_005262472.1 | XP_005262529.1 | 4 | 429 | |||
| XM_017029843.1 | XP_016885332.1 | 4 | 426 | |||
| GenBank mRNA | AK301478.1 | - | 4 | 463 | ||
| AL832120.1 | - | 4 | 366 | |||
| BC040518.1 | - | 4 | 438 | |||
| BC064350.1 | - | 4 | 377 | |||